3-Ethoxy-4-isopropoxybenzaldehyd

Lieferant: FLUOROCHEM

014192-1G 014192-5G
FLUO014192-1GEA 168 EUR
FLUO014192-1G FLUO014192-5G
3-Ethoxy-4-isopropoxybenzaldehyd
3-Ethoxy-4-isopropoxybenzaldehyd
Formel: C₁₂H₁₆O₃
MDL: MFCD01921990
CAS-Nummer: 284044-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR