3-Ethoxy-4-methoxybenzaldehyd ≥99%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Apremilast Impurity 28

M-Clarity™ Qualitätsstufe = MQ100

252751-25G 252751-5G
252751-25GEA 245 EUR
252751-25G 252751-5G
3-Ethoxy-4-methoxybenzaldehyd ≥99%, Sigma-Aldrich®
3-Ethoxy-4-methoxybenzaldehyde

3-Ethoxy-4-methoxybenzaldehyde is a derivative of vanillin. 3-Ethoxy-4-methoxybenzaldehyde may be used to synthesise Compound of Designation red 10 binding (CDr10b) to investigate its role in neuroinflammatory diseases.

Formel: C₁₀H₁₂O₃
Molecular Weight: 180.20 g/mol
Schmelzpunkt: 51...53 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00010128
CAS-Nummer: 1131-52-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2c454a8e65bd06cd919c0e49104f936be6bd23f&vid=a2c454a8e65bd06cd919c0e49104f936be6bd23f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to Tan
Appearance (Form); Powder, Crystals, Crystalline Powder and/or Chunks Conforms to Requirements
Infrared spectrum Conforms to Structure
Purity (GC) ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR