cis-Jasmon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (Z)-3-Methyl-2-(pent-2-en-1-yl)cyclopent-2-enone, cis-Jasmon

M-Clarity™ Qualitätsstufe = MQ300

W319600-100G-K W319600-1KG-K W319600-5KG-K
SIALW319600-100G-KEA 213 EUR
SIALW319600-100G-K SIALW319600-1KG-K SIALW319600-5KG-K
cis-Jasmon, Sigma-Aldrich®
cis-Jasmon
Formel: C₁₁H₁₆O
Molecular Weight: 164.25 g/mol
MDL: MFCD00001402
CAS-Nummer: 488-10-8
EINECS: 207-668-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR