4-(3-Fluorpropoxy)benzaldehyd

Lieferant: BIONET RESEARCH

11P-060.500 11P-060.1 11P-060.5 11P-060.10
BIOT11P-060.500EA 0 EUR
BIOT11P-060.500 BIOT11P-060.1 BIOT11P-060.5 BIOT11P-060.10
4-(3-Fluorpropoxy)benzaldehyd
4-(3-Fluoropropoxy)benzaldehyde
Formel: C₁₀H₁₁FO₂
MDL: MFCD02090999
CAS-Nummer: 400825-68-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR