2-Chlor-6-methoxy-4-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chloro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloro-6-methoxypyridine-4-boronic acid pinacol ester
PN-5547-1G
PN-5547-250M
COBBPN-5547-1GEA
983
EUR
COBBPN-5547-1G
COBBPN-5547-250M
2-Chlor-6-methoxy-4-pyridinboronsäurepinakolester
2-Chlor-6-methoxy-4-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₇BClNO₃ Molecular Weight: 269,5323 g/mol |
MDL:
MFCD11617855 CAS-Nummer: 697739-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...