2-Benzyloxy-5-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
6-Benzyloxypyridine-3-boronic acid pinacol ester, 6-Benzyloxypyridine-3-boronic acid, pinacol ester, 2-(Benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PN-5175-5G
PN-5175-1G
COBBPN-5175-5GEA
503
EUR
COBBPN-5175-5G
COBBPN-5175-1G
2-Benzyloxy-5-pyridinboronsäurepinakolester
2-Benzyloxy-5-pyridinboronsäurepinakolester
Formel:
C₁₈H₂₂BNO₃ Molecular Weight: 311.19 g/mol |
MDL:
MFCD06798263 CAS-Nummer: 832735-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...