Methyl-3-(brommethyl)benzoat

Lieferant: Thermo Scientific

Synonyme: Methyl 3-(bromomethyl)benzoate

TL00214.1 TL00214EA TL00214.50
MAYBTL00214.1EA 0 EUR
MAYBTL00214.1 MAYBTL00214EA MAYBTL00214.50
Methyl-3-(brommethyl)benzoat
Methyl-3-(brommethyl)benzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229,07 g/mol
MDL: MFCD00051437
CAS-Nummer: 1129-28-8
EINECS: 419-100-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR