Chromon-3-carbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A14422.03 A14422.06 A14422.14
A14422.03EA 35.7 EUR
A14422.03 A14422.06 A14422.14
Chromon-3-carbonsäure ≥98%
Chromon-3-carbonsäure

Purity: 98% CAS No.: 39079-62-4 Molecular Formula: C10H6O4 Formula Weight: 190.15 Melting Point: 198-203° Beilstein Reference: 1074111

Formel: C₁₀H₆O₄
Schmelzpunkt: 198…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017338
CAS-Nummer: 39079-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR