N-(Chloracetyl)morpholin

Lieferant: FLUOROCHEM

Synonyme: 4-(Chloroacetyl)morpholine, 4-(2-Chloroacetyl)morpholine

005874-2.5G 005874-25G 005874-5G
FLUO005874-2.5GEA 0 EUR
FLUO005874-2.5G FLUO005874-25G FLUO005874-5G
N-(Chloracetyl)morpholin
N-(Chloracetyl)morpholin
Formel: C₆H₁₀ClNO₂
Molecular Weight: 163.61 g/mol
MDL: MFCD00721939
CAS-Nummer: 1440-61-5
EINECS: 215-880-3
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR