3-(Benzyloxy)-4-methoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Benzyloxy-4-methoxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ200

163953-10G 163953-50G
SIAL163953-10GEA 65.1 EUR
SIAL163953-10G SIAL163953-50G
3-(Benzyloxy)-4-methoxybenzaldehyd, Sigma-Aldrich®
3-(Benzyloxy)-4-methoxybenzaldehyd
Formel: C₁₅H₁₄O₃
Molecular Weight: 242,27 g/mol
MDL: MFCD00003386
CAS-Nummer: 6346-05-0
EINECS: 228-752-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR