2-Hydroxy-6-methylnicotinsäure ≥98%
Lieferant: Thermo Scientific
Synonyme: 2-Hydroxy-6-methylpyridine-3-carboxylic acid
A11842.06 A11842.14
A11842.06EA 22.7 EUR
A11842.06 A11842.14
2-Hydroxy-6-methylnicotinsäure ≥98%
2-Hydroxy-6-methylnicotinsäure
Purity: 98+% CAS No.: 38116-61-9 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 233-238° Beilstein Reference: 129164
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 233…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006272
CAS-Nummer: 38116-61-9
EINECS: 253-784-3