2-Hydroxy-6-methylnicotinsäure

Lieferant: Thermo Scientific

Synonyme: 2-Hydroxy-6-methylpyridine-3-carboxylic acid

SB01990.1 SB01990.50
MAYBSB01990.1EA 0 EUR
MAYBSB01990.1 MAYBSB01990.50
2-Hydroxy-6-methylnicotinsäure
2-Hydroxy-6-methylnicotinsäure
Formel: C₇H₇NO₃
Molecular Weight: 153.14 g/mol
MDL: MFCD00006272
CAS-Nummer: 38116-61-9
EINECS: 253-784-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR