Ethyl-2-oxocyclopentancarboxylat

Lieferant: CHEMPUR

Synonyme: Ethylcyclopentanone-2-carboxylate, Ethyl 2-oxocyclopentanecarboxylate

JFD04002.250 JFD04002.1
CHMPJFD04002.250EA 0 EUR
CHMPJFD04002.250 MAYBJFD04002.1
Ethyl-2-oxocyclopentancarboxylat
Ethyl-2-oxocyclopentancarboxylat
Formel: C₈H₁₂O₃
Molecular Weight: 156,18 g/mol
MDL: MFCD00001412
CAS-Nummer: 611-10-9
EINECS: 210-253-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR