Ethyl-2-oxocyclopentancarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethylcyclopentanone-2-carboxylate, Ethyl 2-oxocyclopentanecarboxylate

M-Clarity™ Qualitätsstufe = MQ200

168092-100G 168092-25G
SIAL168092-100GEA 158 EUR
SIAL168092-100G SIAL168092-25G
Ethyl-2-oxocyclopentancarboxylat, Sigma-Aldrich®
Ethyl-2-oxocyclopentancarboxylat
Formel: C₈H₁₂O₃
Molecular Weight: 156,18 g/mol
MDL: MFCD00001412
CAS-Nummer: 611-10-9
EINECS: 210-253-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR