3-(Brommethyl)-5-chlorbenzo[b]thiophen

Lieferant: FLUOROCHEM

CFF343.2.5 CFF343.10 017379-2.5G 017379-10G 017379-1G 017379-5G
CHMPCFF343.2.5EA 481 EUR
CHMPCFF343.2.5 CHMPCFF343.10 FLUO017379-2.5G FLUO017379-10G FLUO017379-1G FLUO017379-5G
3-(Brommethyl)-5-chlorbenzo[b]thiophen
3-(Brommethyl)-5-chlorbenzo[b]thiophen
Formel: C₉H₆BrClS
Molecular Weight: 261,5700 g/mol
MDL: MFCD00173769
CAS-Nummer: 1198-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR