3-(Brommethyl)-5-chlorbenzo[b]thiophen
Lieferant: Thermo Scientific
MWP00055.250
MWP00055DA
MAYBMWP00055.250EA
0
EUR
MAYBMWP00055.250
MAYBMWP00055DA
3-(Brommethyl)-5-chlorbenzo[b]thiophen
3-(Brommethyl)-5-chlorbenzo[b]thiophen
Formel:
C₉H₆BrClS Molecular Weight: 261,5700 g/mol |
MDL:
MFCD00173769 CAS-Nummer: 1198-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...