Harnesses with positioning belt, Titan™
Lieferant: HONEYWELL SAFETY
1011894 1011893
113-1270EA 144 EUR
113-1270 DALL1011893
Harnesses with positioning belt, Titan™
Fallschutz Auffanggurte
Lightweight harnesses for building and construction work. Available as a 1-point version with rear anchorage or as a 2-point version with front and rear anchorage and front webbing loops.
  • Lightweight, durable polyamide webbing for all day comfort and dependability
  • Fully adjustable designs so one size fits all
  • Two lateral D-rings (Dorsal and side)
  • Sub pelvic strap keeps the wearer upright
  • Belt and fast release buckles
EN 361
Standards: EN 361