Anti-alpha-Actin (1A4) Mouse Monoclonal Antibody

Lieferant: Merck Millipore (Oncogene)
113200-500
113200-500EA 583 EUR
113200-500
Anti-alpha-Actin (1A4) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: alpha-Actin (1A4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR