7-Aminoactinomycin (7-AAD)-Lösung Verwendung in der nuklearen Färbung

Lieferant: Tonbo Biosciences

13-6993-T500 13-6993-T200
13-6993-T500EA 40.5 EUR
13-6993-T500 13-6993-T200
7-Aminoactinomycin (7-AAD)-Lösung Verwendung in der nuklearen Färbung
7-Aminoactinomycin D (7-AAD)

This nucleic acid dye can be used to exclude non viable cells from the analysis of flow cytometry data.


Useful as a viability probe for exclusion of non viable cells based on light scatter and fluorescence properties.


Bitte beachten Protect skin and eyes by wearing suitable protective clothing, gloves and eye/face protection.

Formel: C₆₂H₈₇N₁₃O₁₆
MDL: MFCD00077295
CAS-Nummer: 7240-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR