Chromatography Tubing, PEEK
Lieferant: Avantor
16-PEEK-05-10MEA 341 EUR
16-PEEK-05-10M 16-PEEK-07-10M 16-PEEK-07S-10 16-PEEK-10-10M 16-PEEK-20-10M 16-PEEK-02-3M 16-PEEK-03-3M 16-PEEK-04-3M 16-PEEK-05-3M 16-PEEK-07-3M 16-PEEK-07S-3M 16-PEEK-10-3M 16-PEEK-20-3M 16-PEEK-30-3M 16-PEEK-02-10M 16-PEEK-03-10M 16-PEEK-04-10M 16-PEEK-30-10M 16-PEEK-03-5M 16-PEEK-04-5M 16-PEEK-05-5M 16-PEEK-07-5M 16-PEEK-10-5M 16-PEEK-20-5M 16-PEEK-30-5M 16-PEEK-20-25M
Chromatography Tubing, PEEK
Schläuche Chromatographie-Schläuche
PEEK
  • Inert and biocompatible
  • Excellent for high pressure applications (413 bar)
Ten times less oxygen permeable than PTFE and with a greater temperature resistance. Completely flexible, easy to cut and suitable for a wide range of applications. Can be used with PEEK fingertight fittings or stainless steel nuts and ferrules.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable