Anti-APOE Goat Polyclonal Antibody

Lieferant: Merck Millipore (Oncogene)
178479-500
178479-500EA 500 EUR
178479-500
Anti-APOE Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR