CF™594 antibodies are affinity-purified antibodies labeled with deep red fluorescent CF™594, a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™594 is a deep red fluorescent dye with excitation/emission maxima at 593/614nm. CF™594 is spectrally similar to AlexaFluor®594, ATTO 594, DyLight®594, and Texas Red®. CF™594 protein conjugates are significantly brighter than those of AlexaFluor®594. CF™594 is also highly photostable, a critical feature for applications such as confocal microscopy. The deep red fluorescence emission of CF™594 has minimal spill-over in the green channel, making it an excellent partner for green fluorescent CF™488A for multicolor imaging.
CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor® and Texas Red® are registered trademarks of Molecular Probes, Inc. DyLight® is a registered trademark of Pierce Biotechnology.
Bestellinformation: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.
Type: Secondary
Antigen: IgM MU CHAIN
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human