Anti-IgG Chicken Polyclonal Antibody (CF™ 350)

Lieferant: Biotium
20364-50UL
20364-50ULEA 121 EUR
20364-50UL
Anti-IgG Chicken Polyclonal Antibody (CF™ 350)
Antikörper
CF™350 is a blue fluorescent dye with excitation/emission maxima at 347/448nm. CF™350 is optimized for UV excitation, and is spectrally similar to AlexaFluor®350, AMCA, and DyLight®350. CF™350 is more water soluble, more photostable, and 50% brighter when conjugated to antibodies than the traditional blue dye AMCA. CF™350 antibody conjugates are comparable in brightness to AlexaFluor®350 dye conjugates.

CF™350 is a blue fluorescent dye with excitation/emission maxima at347/448 nm. CF™350 is optimized for UV excitation, and is spectrally similar to AlexaFluor®350, AMCA, and DyLight™350. CF™350 is more water soluble, more photostable, and 50% brighter when conjugated to antibodies than the traditional blue dye AMCA. CF™350 antibody conjugates are comparable in brightness to AlexaFluor®350 dye conjugates.

CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor® is a registered trademark of Molecular Probes, Inc. DyLight® is a registered trademark of Pierce Biotechnology.

Bestellinformation: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF350
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR