Anti-IgG Goat Polyclonal Antibody (CF™ 532)

Lieferant: Biotium
20365-500UL
20365-500ULEA 300 EUR
20365-500UL
Anti-IgG Goat Polyclonal Antibody (CF™ 532)
Antikörper
CF™532 antibodies are affinity-purified antibodies labeled with orange fluorescent CF™532 (Ex/Em: 527/558nm), a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.

CF™532 is an orange fluorescent dye with an excitation spectrum optimally matching the 532nm line of the frequency-doubled Nd-Yag laser. Antibody conjugates of the dye are much brighter than those of AlexaFluor® 532.

CF™ dyes are covered by pending U.S. and international patents.

Bestellinformation: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF532
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR