Anti-IgG Donkey Polyclonal Antibody (CF™ 405S)
Lieferant: Biotium
20416-1MG
20416-1MGEA 368 EUR
20416-1MG
Anti-IgG Donkey Polyclonal Antibody (CF™ 405S)
Antikörper
This product is prepared by labeling highly cross-adsorbed donkey anti-goat IgG (H+L) with CF™405S. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S dye results in much brighter signal than that using Alexa Fluor® 405. To minimize cross reactivity, the antibody has been adsorbed against chicken, guinea pig, Syrian hamster, horse, human, mouse, rabbit, and rat serum proteins.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF405S
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat