Anti-GST Goat Polyclonal Antibody (CF594)

Lieferant: Biotium
20425
20425.EA 436 EUR
20425.
Anti-GST Goat Polyclonal Antibody (CF594)
Antikörper
Polyclonal anti-GST reacts with glutathione-S transferase (GST), a common expression tag used in molecular biology. This conjugate is prepared by labeling polyclonal goat anti-GST IgG (H+L) with CF™594 dye. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Tag
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Goat
Isotype:
Reactivity: GST
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR