Anti-GST Goat Polyclonal Antibody (CF640R)

Lieferant: Biotium
20426
20426.EA 436 EUR
20426.
Anti-GST Goat Polyclonal Antibody (CF640R)
Antikörper
Polyclonal anti-GST reacts with glutathione-S transferase (GST), a common expression tag used in molecular biology. This conjugate is prepared by labeling polyclonal goat anti-GST IgG (H+L) with CF™640R dye. CF™640R (Ex/Em 642/662 nm) is a novel far-red rhodamine-based dye spectrally similar to Cy®5 and Alexa Fluor® 647. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635 nm. However, the most important advantage of CF™640R is its exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Tag
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Goat
Isotype:
Reactivity: GST
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR