Anti-Myc Tag Rabbit Polyclonal Antibody (CF640R)

Lieferant: Biotium
20439
20439.EA 325 EUR
20439.
Anti-Myc Tag Rabbit Polyclonal Antibody (CF640R)
Antikörper
Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-MYC IgG (H+L) with CF™640R dye. CF™640R (Ex/Em 642/662 nm) is a novel far-red rhodamine-based dye spectrally similar to Cy®5 and Alexa Fluor® 647. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635 nm. However, the most important advantage of CF™640R is its exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Tag
Antigen: Myc Tag
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Rabbit
Isotype:
Reactivity: MYC tag
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR