Goat anti-alpaca IgG antibody that recognizes the VHH domain of heavy chain alpaca IgG. Labeled with CF® Dyes and suitable for western or IF applications.
- Available in 6 bright and photostable CF® Dyes
- Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
- CF® Dye and biotin conjugates: add 0.5 mL distilled water
- HRP or DNP conjugates: add 1 ml distilled water
Goat Anti-Alpaca IgG, VHH antibody is an affinity-purified polyclonal goat IgG whole antibody that recognizes the VHH domain of heavy chain alpaca IgG, subclasses 2 and 3, and the VHH domain of llama IgG, subclasses 2 and 3. The antibody is cross-adsorbed against bovine, human, mouse, rabbit, and rat serum proteins. It is not highly cross-adsorbed against immunoglobulins from other species. The antibody is available conjugated to Biotium’s bright and photostable CF® Dyes.
Type: Secondary
Antigen: Alpaca
Clonality: Polyclonal
Clone:
Conjugation: CF®647
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Alpaca, Llama