Anti-Dopamine D 2 Receptor Rabbit Polyclonal Antibody

Lieferant: Merck Millipore (Oncogene)
324393-100
324393-100EA 583 EUR
324393-100
Anti-Dopamine D 2 Receptor Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Dopamine D 2 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR