Anti-Ganglioside Rabbit Polyclonal Antibody

Lieferant: Merck Millipore (Oncogene)
345758-100
345758-100EA 450 EUR
345758-100
Anti-Ganglioside Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Ganglioside
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR