Anti-G i alpha-1 and Rabbit Polyclonal Antibody
Lieferant: Merck Millipore (Oncogene)
371723-50
371723-50EA 614 EUR
371723-50
Anti-G i alpha-1 and Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: G i alpha-1 and
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen