Peroxidase Indicator Reagent
Lieferant: SIGMA ALDRICH MICROSCOPY
3901-10VL
3901-10VLEA
379
EUR
3901-10VL
Peroxidase Indicator Reagent
Elektrophorese-Reagenzien
Western Blotting Reagents
It contains p-Phenylenediamine diHCL (1 part) and catechol (2 parts). Commonly referred to as Hanker Yates reagent. Kit component in the Sigma Leukocyte peroxidase (myeloperoxidase) Procedure, 390A.