Anti-IP 3 Receptor (IPR.1) Mouse Monoclonal Antibody

Lieferant: Merck Millipore (Oncogene)
407140-50
407140-50EA 600 EUR
407140-50
Anti-IP 3 Receptor (IPR.1) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: IP 3 Receptor (IPR.1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR