Ultrasonic processors, Vibra-Cell™ VCX 500, VCX 750, VCX 850 and VCX 1500
Lieferant: SONICS MATERIALS
VCX750 VCX500 VCX1500
432-0138EA 8680 EUR
432-0138 432-0137 432-0139
Ultrasonic processors, Vibra-Cell™ VCX 500, VCX 750, VCX 850 and VCX 1500
Homogenisatoren Ultraschall-Desintegratoren
The Vibra-Cell™ is a technologically advanced, high-intensity ultrasonic processor for high and low volume applications*. It can safely process a wide variety of organic and inorganic materials in applications such as cell disruption, sample preparation, homogenisation, disaggregation and sonochemical reactions. The system can vary the power output and constantly monitor both power (Watts) and energy (Joules). The microprocessor control with real time display gives digital accuracy and reproducible control of set and run parameters.
  • Automatic tuning eliminates need for constant
More Product Information