Anti-Mannose 6-Phosphate Receptor (2G11) Mouse Monoclonal Antibody

Lieferant: Merck Millipore (Oncogene)
444105-100
444105-100EA 541 EUR
444105-100
Anti-Mannose 6-Phosphate Receptor (2G11) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Mannose 6-Phosphate Receptor (2G11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR