Anti-Neutrophil Elastase Rabbit Polyclonal Antibody
Lieferant: Merck Millipore (Oncogene)
481001-1
481001-1EA 208 EUR
481001-1
Anti-Neutrophil Elastase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Neutrophil Elastase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human