Anti-Phosphotyrosine (PY20) Mouse Monoclonal Antibody [clone: PY20] Peroxidase

Lieferant: Merck Millipore (Oncogene)
525320-100
525320-100EA 555 EUR
525320-100
Anti-Phosphotyrosine (PY20) Mouse Monoclonal Antibody [clone: PY20] Peroxidase
Antikörper
Type: Primary
Antigen: Phosphotyrosine (PY20)
Clonality: monoclonal
Clone:
Conjugation: Peroxidase
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR