Anti-PP2A/Balpha (14-27) (PR55) Rabbit Polyclonal Antibody
Lieferant: Merck Millipore (Oncogene)
539521-100
539521-100EA 640 EUR
539521-100
Anti-PP2A/Balpha (14-27) (PR55) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PP2A/Balpha (14-27) (PR55)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen