Anti-MAPT Rabbit Polyclonal Antibody

Lieferant: Merck Millipore (Oncogene)
577815-10
577815-10EA 612 EUR
577815-10
Anti-MAPT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR