Anti-Tissue Factor (TF9-10H10) Mouse Monoclonal Antibody [clone: TF9-10H10]
Lieferant: Merck Millipore (Oncogene)
612161-500
612161-500EA 569 EUR
612161-500
Anti-Tissue Factor (TF9-10H10) Mouse Monoclonal Antibody [clone: TF9-10H10]
Antikörper
Type: Primary
Antigen: Tissue Factor (TF9-10H10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen