Vector, pET-24d(+) DNA
Lieferant: Merck Millipore (Novagen)
69752-3
69752-3EA 291 EUR
69752-3
Vector, pET-24d(+) DNA
Vektoren, Plasmide und Bibliotheken
The pET-24a-d(+) vectors carry an N-terminal T7·Tag® sequence plus an optional C-terminal His·Tag® sequence.
Lieferumfang: The pET vectors are supplied as purified plasmid DNA (10 µg). And also includes an induction control strain (supplied as a glycerol stock).