His·Bind® buffer kit, Novagen®
Lieferant: Merck Millipore (Novagen)
69755-3
69755-3EA 303 EUR
69755-3
His·Bind® buffer kit, Novagen®
Puffer Puffer für die Antikörperreinigung
The His·Bind buffer kit is a set of pre-tested buffers designed for use with IDA-based His·Bind resin for convenient, rapid one-step purification of proteins by metal chelation chromatography.
Solutions are included for Ni²⁺ charging, binding, washing and elution of up to ten 2,5 ml columns.
Lieferumfang: The kit contains 8× binding buffer (2×80 ml), 8× wash buffer (25 ml), 4× elute buffer (50 ml), 4× strip buffer (50 ml) and 8× charge buffer (20 ml).