Vector, pET-24(+) DNA
Lieferant: Merck Millipore (Novagen)
69772-3
69772-3EA
306
EUR
69772-3
Vector, pET-24(+) DNA
Vektoren, Plasmide und Bibliotheken
pET-24(+) is a transcription vector designed for expression from bacterial translation signals carried within a cloned insert. It lacks the ribosome binding site and ATG start codon present on the pET translation vectors. A C-terminal His·Tag® sequence is available
Lieferumfang: pET vectors are supplied as purified plasmid DNA (10 µg) and an induction control strain (supplied as a glycerol stock).