IPTG (Isopropyl-β-D-thiogalactopyranosid)-Lösung 100 mM, Sigma-Aldrich®

Lieferant: Merck Millipore (Novagen)

Synonyme: IPTG

70527-3
70527-3EA 137 EUR
70527-3
IPTG (Isopropyl-β-D-thiogalactopyranosid)-Lösung 100 mM, Sigma-Aldrich®
IPTG (Isopropyl-β-D-thiogalactopyranosid)

The lac operon inducer IPTG (isopropyl-β-D- 1-thiogalactopyranoside) is recommended for blue/white screening by lacZ α- complementation with appropriate vectors and host strains, such as pSTBlue-1/NovaBlue, and for inducing protein expression with the pET System and other lac promoter-derived expression systems.

Formel: C₉H₁₈O₅S
Molecular Weight: 238,31 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1

Bestellen

Spezifikation Testergebnisse

Storage -20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR