Protein extraction reagent, BugBuster® HT, Millipore®
Lieferant: Merck Millipore (Novagen)
70922-3 70922-5 70922-4
70922-3EA 240 EUR
70922-3 70922-5 70922-4
Protein extraction reagent, BugBuster® HT, Millipore®
Puffer Extraktionspuffer
BugBuster® HT Protein Extraction Reagent combines BugBuster® Protein Extraction Reagent and Benzonase® Nuclease in one convenient reagent. BugBuster® HT eliminates common bioprocessing problems resulting from traditional cell lysis procedures. Soluble proteins are gently extracted from E. coli without exposure to heat or oxidative damage and viscosity is eliminated by nucleic acid digestion in a single step. The resulting protein extract can easily be fractionated by conventional purification techniques. BugBuster HT is ideally suited for application in high throughput protein purifications.
Storage: 2 to 8 °C