Protein extraction reagent, BugBuster® (primary amine-free), Millipore®
Lieferant: Merck Millipore (Novagen)
BugBuster® (primary amine-free) Protein Extraction Reagent is a special formulation of BugBuster designed for applications where primary amines would interfere if present in the protein extract, such as protein immobilisation or cross-linking. The PIPPS buffer used in BugBuster® (primary amine-free) has a similar buffer capacity and pH range as the original Tris-buffered BugBuster®, but will not complex metal ions, also making it ideally suited for extraction of metal dependent proteins.
Storage: 15 to 30 °C