Electroporation solution, Ingenio®
Lieferant: MIRUS
MIR50111 MIR50114 MIR50117
732-1215EA 341 EUR
732-1215 732-1216 732-1217
Electroporation solution, Ingenio®
Nukleinsäure-Reagenzien Transformations-Reagenzien
Ingenio® Electroporation solution is compatible with many conventional electroporation instruments, including Lonza Amaxa® Nucleofector®, Bio-Rad® Gene Pulser and Harvard-BTX® electroporators. It can be used for electroporation with both exponential decay as well as square wave forms.