Precast gels, Latitude™ HT Gels and Midigels
Lieferant: LONZA
57220 57226 57246 57224 57230 57231 57234
733-1740EA 328 EUR
733-1740 733-1741 733-1743 733-1596 733-1598 733-1599 733-1601
Precast gels, Latitude™ HT Gels and Midigels
Elektrophorese-Gele Fertiggele
Latitude™ DNA gels are designed for accurate DNA fragment separation for almost any gel electrophoresis application. These ready-to-run gels are available in a variety of sizes and well formats to fit most high-throughput needs.
  • Made with high quality, molecular biology grade SeaKem® or NuSieve® agarose
  • DNase- and RNase-free
  • Provide sharp bands with low background fluorescence
  • Ideal for checking restriction digests and PCR products
  • Fit in most large submerged electrophoresis systems and are compatible with multichannel pipettors
Both the DNA HT gelsMore Product Information