Stainless steel manipulator, Corning® HYPERStack®
Lieferant: Corning
11000
734-2491EA 24430 EUR
734-2491
Stainless steel manipulator, Corning® HYPERStack®
Kolben Zellkulturkolben
The Corning® HYPERStack™ Stainless Steel Manipulator accommodates both HYPERStack™-36 and CellSTACK-10 vessels.
It handles two units of the same type simultaneously or one unit at a time. It is ergonomically designed and has large, easy to read dials that are clearly marked to identify positions for ease of use in protocol development.